missing translation for 'onlineSavingsMsg'
Learn More

Dihydroergotamine mesylate, MedChemExpress

Product Code. 16498568
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498568 10 mg 10mg
16414447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498568 Supplier Medchem Express Supplier No. HYB0670A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dihydroergotamine mesylate is an ergot alkaloid used to treat migraines.

Dihydroergotamine mesylate is an ergot alkaloid used to treat migraines.

Specifications

Chemical Name or Material Dihydroergotamine mesylate
CAS 6190-39-2
Color White
Purity Grade Notes Research
Molecular Formula C34H41N5O8S
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (73.55 mM; Need ultrasonic)
SMILES O[C@@]([C@@](CCC1)([H])N1C2=O)(O[C@](NC([C@@H](CN(C)[C@]3([H])C4)C[C@]3([H])C5=C6C4=CNC6=CC=C5)=O)(C)C7=O)N7[C@H]2CC8=CC=CC=C8.CS(=O)(O)=O
Molecular Weight (g/mol) 679.78
Formula Weight 679.78
Percent Purity 98.42%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.