missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydroergotamine Mesilate, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Dihydroergotamine mesilate |
| CAS | 6190-39-2 |
| Analyte or Component Names | Dihydroergotamine mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H37 N5 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dihydroergotamine mesilate, (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-methyl-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a- octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate, Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-methyl-5'-(phenylmethyl)-, (5'α,10α)-, monomethanesulfonate (salt) (9CI), Ergotamine, 9,10-dihydro-, monomethanesulfonate (salt) (6CI,7CI,8CI), (+)-Dihydroergotamine mesylate, 9,10-Dihydroergotamine mesylate, Agit, Angionorm, DETMS, DHE 45, Dergotamine, Diergo, Dihydergot, Dihydroergotamine mesylate, Dihydroergotamine methanesulfonate, Dihydroergotamine methanesulphonate, Dihydroergotamine monomethanesulfonate, Dihytamine, Dirgotarl, Endophleban, Ergomimet, Ergont, Ergotonin, Ikaran, Migranal, Morena, Orstanorm, Seglor, Tonopres, Verladyn |
| InChI Formula | InChI=1S/C33H37N5O5.CH4O3S/c1-32(35-29(39)21-15-23-22-10-6-11-24-28(22)20(17-34-24)16-25(23)36(2)18-21)31(41)38-26(14-19-8-4-3-5-9-19)30(40)37-13-7-12-27(37)33(38,42)43-32;1-5(2,3)4/h3-6,8-11,17,21,23,25-27,34,42H,7,12-16,18H2,1-2H3,(H,35,39);1H3,(H,2,3,4)/t21-,23-,25-,26+,27+,32-,33+;/m1./s1 |
| Solution Type | Neat |
| SMILES | CN1C[C@@H](C[C@H]2[C@H]1Cc3c[nH]c4cccc2c34)C(=O)N[C@]5(C)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C5=O.CS(=O)(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction