missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihydroergocristine Mesilate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Dihydroergocristine Mesilate
Specifications
Specifications
| Chemical Name or Material | Dihydroergocristine Mesilate |
| CAS | 24730-10-7 |
| Analyte or Component Names | Dihydroergocristine Mesilate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H41 N5 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dihydroergocristine Mesilate, (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-(1-methylethyl)-3,6-dioxo-octahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate, Ergocristine, 9,10-dihydro-, monomethanesulfonate (salt) (8CI), Ergocristine, dihydro-, methanesulfonate (6CI,7CI), 9,10-Dihydroergocristine methanesulfonate, Dihydroergocristine mesylate, Dihydroergocristine methanesulfonate, Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(phenylmethyl)-, (5'alpha,10alpha)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 9,10-dihydro-12'-hydroxy-2'-(1-methylethyl)-5'-(phenylmethyl)-, (5'alpha,10alpha)-, monomethanesulfonate (salt) (9CI), Dihydroergotamine Mesilate Impurity E as Mesilate, Dihydroergotamine Impurity E as Mesilate |
| InChI Formula | InChI=1S/C35H41N5O5.CH4O3S/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34;1-5(2,3)4/h4-7,9-12,18,20,23,25,27-29,36,44H,8,13-17,19H2,1-3H3,(H,37,41);1H3,(H,2,3,4)/t23-,25-,27-,28+,29+,34-,35+;/m1./s1 |
| SMILES | CC(C)[C@@]1(NC(=O)[C@@H]2C[C@H]3[C@@H](Cc4c[nH]c5cccc3c45)N(C)C2)O[C@@]6(O)[C@@H]7CCCN7C(=O)[C@H](Cc8ccccc8)N6C1=O.CS(=O)(=O)O |
| IUPAC Name | (6aR,9R,10aR)-N-[(2R,5S,10aS,10bS)-5-Benzyl-10b-hydroxy-2-(1-methylethyl)-3,6-dioxo-octahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction