missing translation for 'onlineSavingsMsg'
Learn More

Dihydrodaidzein, MedChemExpress

Product Code. 16439228
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16439228

Brand: Medchem Express HYN1461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dihydrodaidzein is one of the most prominent dietary phytoestrogens.

Dihydrodaidzein is one of the most prominent dietary phytoestrogens.

Specifications

Chemical Name or Material Dihydrodaidzein
CAS 17238-05-0
Color Gray
Purity Grade Notes Research
Molecular Formula C15H12O4
Quantity 10 mg
Synonym (±)-Dihydrodaidzein
Solubility Information DMSO : 100 mg/mL (390.24 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(O)C=C2)COC3=CC(O)=CC=C13
Molecular Weight (g/mol) 256.25
Formula Weight 256.25
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.