missing translation for 'onlineSavingsMsg'
Learn More

Dihydrodaidzein, MedChemExpress

Product Code. 16471638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471638 5 mg 5mg
16439228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471638 Supplier Medchem Express Supplier No. HYN1461
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dihydrodaidzein is one of the most prominent dietary phytoestrogens.

Dihydrodaidzein is one of the most prominent dietary phytoestrogens.

Specifications

Chemical Name or Material Dihydrodaidzein
CAS 17238-05-0
Color Gray
Purity Grade Notes Research
Molecular Formula C15H12O4
Quantity 5 mg
Synonym (±)-Dihydrodaidzein
Solubility Information DMSO : 100 mg/mL (390.24 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(O)C=C2)COC3=CC(O)=CC=C13
Molecular Weight (g/mol) 256.25
Formula Weight 256.25
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.