missing translation for 'onlineSavingsMsg'
Learn More

Dihydrocoumarin, MedChemExpress

Product Code. 16499166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499166 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499166 Supplier Medchem Express Supplier No. HYN1926
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Dihydrocoumarin is a compound found in Melilotus officinalis. Dihydrocoumarin is a yeast Sir2p inhibitor. Dihydrocoumarin also inhibits human SIRT1 and SIRT2 with IC50s of 208 μM and 295 μM, respectively.

Dihydrocoumarin is a compound found in Melilotus officinalis. Dihydrocoumarin is a yeast Sir2p inhibitor. Dihydrocoumarin also inhibits human SIRT1 and SIRT2 with IC50s of 208 μM and 295 μM, respectively.

Specifications

Chemical Name or Material Dihydrocoumarin
CAS 119-84-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H8O2
Quantity 100 mg
Synonym Hydrocoumarin Chroman-2-one
Solubility Information DMSO : 100 mg/mL (674.95 mM; Need ultrasonic)
SMILES O=C1CCC2=CC=CC=C2O1
Molecular Weight (g/mol) 148.16
Formula Weight 148.16
Percent Purity 99.09%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.