missing translation for 'onlineSavingsMsg'
Learn More

Dihydrocoumarin, 99%

Product Code. 11424747
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424747 2500 g 2500g
11404747 100 g 100g
11414747 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424747 Supplier Thermo Scientific Alfa Aesar Supplier No. B21229.0E
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-84-6 | C9H8O2 | 148.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-84-6
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006881
InChI Key VMUXSMXIQBNMGZ-UHFFFAOYSA-N
Synonym dihydrocoumarin, 3,4-dihydrocoumarin, hydrocoumarin, chroman-2-one, benzodihydropyrone, melilotin, melilotol, 2-chromanone, 1,2-benzodihydropyrone, melilotic lactone
PubChem CID 660
ChEBI CHEBI:16151
IUPAC Name 3,4-dihydrochromen-2-one
SMILES C1CC(=O)OC2=CC=CC=C21

Specifications

Melting Point 24°C to 25°C
Density 1.169
Boiling Point 272°C
Flash Point 130°C (266°F)
Refractive Index 1.556
Quantity 2500 g
Beilstein 4584
Formula Weight 148.16
Percent Purity 99%
Chemical Name or Material Dihydrocoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.