missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dihydrocarvone, 98%, mixture of isomers

Product Code. 10042660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042660 25 g Glass bottle 25g
10388290 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10042660 Supplier Thermo Scientific Acros Supplier No. 203650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5524-05-0 | C10H16O | 152.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5524-05-0
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00001636
InChI Key AZOCECCLWFDTAP-RKDXNWHRSA-N
Synonym +-dihydrocarvone, d-dihydrocarvone, 1r,4r-dihydrocarvone, e-dihydrocarvone, unii-vta43h364z, 2r-trans-2-methyl-5-1-methylvinyl cyclohexan-1-one, 2r,5r-2-methyl-5-isopropenylcyclohexanone, 2r,5r-5-isopropenyl-2-methylcyclohexanone, 2r,5r-2-methyl-5-prop-1-en-2-yl cyclohexanone, cyclohexanone, 2-methyl-5-1-methylethenyl-, 2r,5r
PubChem CID 22227
ChEBI CHEBI:154
IUPAC Name (2R,5R)-2-methyl-5-prop-1-en-2-ylcyclohexan-1-one
SMILES CC1CCC(CC1=O)C(=C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9255g/mL
Boiling Point 87.0°C to 88.0°C (6.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4700 to 1.4740
Quantity 25 g
Specific Gravity 0.9255
Specific Rotation Condition + 14.00 (25.00°C c=neat)
Specific Rotation + 14.00
Formula Weight 152.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (+)-Dihydrocarvone, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.