missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Montelukast Sodium, TRC

Product Code. 30009162
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009162 10 mg 10mg
30005906 5 mg 5mg
30010005 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009162 Supplier Toronto Research Chemicals Supplier No. TRCD44921510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142147-98-6
Molecular Formula C35 H38 Cl N O3 S . Na
Molecular Weight (g/mol) 610.18
Synonym Cyclopropaneacetic acid, 1-[[[1-[3-[2-(7-chloro-2-quinolinyl)ethyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, monosodium salt, (R)-, Dihydro Montelukast Sodium Salt
IUPAC Name sodium;2-[1-[[(1R)-1-[3-[2-(7-chloroquinolin-2-yl)ethyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetate
SMILES [Na+].CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)[O-])CC2)c3cccc(CCc4ccc5ccc(Cl)cc5n4)c3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C35H38ClNO3S.Na/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;/h3-9,11-12,14,16,20-21,32,40H,10,13,15,17-19,22-23H2,1-2H3,(H,38,39);/q;+1/p-1/t32-;/m1./s1
Formula Weight 609.208
Chemical Name or Material Dihydro Montelukast Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.