missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Ferulic Acid, TRC

Product Code. 30376180
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376180 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376180 Supplier Toronto Research Chemicals Supplier No. TRCD4489001G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroferulic Acid
CAS 1135-23-5
Molecular Formula C10 H12 O4
Quantity 1 g
Synonym Benzenepropanoic acid, 4-hydroxy-3-methoxy-, Hydrocinnamic acid, 4-hydroxy-3-methoxy- (6CI,7CI,8CI), 4-Hydroxy-3-methoxybenzenepropanoic acid, (4-Hydroxy-3-methoxyphenyl)propionic acid, 3-(3-Methoxy-4-hydroxyphenyl)propanoic acid, 3-(3-Methoxy-4-hydroxyphenyl)propionic acid, 3-(3'-Methoxy-4'-hydroxyphenyl)propionic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid, 3-(4-Hydroxy-3-methoxyphenyl)propionic acid, 3-Methoxy-4-hydroxyphenylpropionic acid, 3-Methoxyphloretic acid, Dihydroconiferylic acid, Dihydroferulic acid, Ferulic acid, α,β-dihydro-, Hydroferulic acid, Shorbic acid, β-(4-Hydroxy-3-methoxyphenyl)propionic acid, β-3-Methoxy-4-hydroxyphenylpropionic acid
InChI Formula InChI=1S/C10H12O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2,4,6,11H,3,5H2,1H3,(H,12,13)
SMILES COc1cc(CCC(=O)O)ccc1O
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol) 196.2
Formula Weight 196.0736
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.