missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Daidzein, TRC

Product Code. 30377509
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377509 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377509 Supplier Toronto Research Chemicals Supplier No. TRCD44900025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dihydrodaidzein
CAS 17238-05-0
Purity Grade Notes HPLC
Molecular Formula C15 H12 O4
Quantity 25 mg
InChI Formula InChI=1S/C15H12O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-7,13,16-17H,8H2
SMILES Oc1ccc(cc1)C2COc3cc(O)ccc3C2=O
IUPAC Name 7-hydroxy-3-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
Molecular Weight (g/mol) 256.25
Formula Weight 256.0736
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.