missing translation for 'onlineSavingsMsg'
Learn More

Dihydro Caffeic Acid 3-O-Sulfate Sodium Salt, TRC

Product Code. 30007789
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007789 2.5 mg 2.5mg
30012839 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007789 Supplier Toronto Research Chemicals Supplier No. TRCD4487102.5MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1187945-70-5
Molecular Formula C9H8Na2O7S
Molecular Weight (g/mol) 306.2
Synonym 4-Hydroxy-3-(sulfooxy)benzenepropanoic Acid Sodium Salt;
IUPAC Name disodium;3-(4-hydroxy-3-sulfonatooxyphenyl)propanoate
SMILES C1=CC(=C(C=C1CCC(=O)[O-])OS(=O)(=O)[O-])O.[Na+].[Na+]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C9H10O7S.2Na/c10-7-3-1-6(2-4-9(11)12)5-8(7)16-17(13,14)15;;/h1,3,5,10H,2,4H2,(H,11,12)(H,13,14,15);;/q;2*+1/p-2
Formula Weight 305.979
Percent Purity >95%
Chemical Name or Material Dihydro Caffeic Acid 3-O-Sulfate Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.