missing translation for 'onlineSavingsMsg'
Learn More

Dihydro-Axitinib, TRC

Product Code. 30387285
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387285 25 mg
30376866 10 mg
30375675 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387285 Supplier Toronto Research Chemicals Supplier No. TRCD44835925MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1443118-73-7
Molecular Formula C22H20N4OS
Molecular Weight (g/mol) 388.49
Synonym 2-((1-Acetyl-1H-indazol-6-yl)sulfinyl)-N-methylbenzamide, Axitinib Impurity F
SMILES CNC(=O)c1ccccc1Sc2ccc3c(CCc4ccccn4)n[nH]c3c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H20N4OS/c1-23-22(27)18-7-2-3-8-21(18)28-16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-8,10-11,13-14H,9,12H2,1H3,(H,23,27)(H,25,26)
Chemical Name or Material Dihydro-Axitinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.