missing translation for 'onlineSavingsMsg'
Learn More

Dihydridotetrakis(triphenylphosphine)ruthenium(II), 97%

Product Code. 10612233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10612233 5 g Glass bottle 5g
10578843 500 mg Glass bottle 500mg
10129801 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612233 Supplier Thermo Scientific Acros Supplier No. 367490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19529-00-1 | C72H60P4Ru | 1150.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19529-00-1
Molecular Formula C72H60P4Ru
Molecular Weight (g/mol) 1150.24
MDL Number MFCD04126429
InChI Key BFPFOLJFUVTHEP-UHFFFAOYSA-N
Synonym dihydridotetrakis triphenylphosphine ruthenium ii, tetrakis triphenylphosphine ruthenium dihydride, c6h5 3p 4ruh2, ruthenium,dihydrotetrakis triphenylphosphine, dihydridotetrakis triphenylphosphine ruthenium, tetrakis triphenylphosphine ruthenium
PubChem CID 11981970
IUPAC Name ruthenium;triphenylphosphane
SMILES [Ru].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 181.0°C to 183.0°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Ru)
Packaging Glass bottle
Linear Formula [(C6H5)3P]4RuH2
Quantity 5 g
Formula Weight 1152.25
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Show More Show Less
compliance-icons
Product Identifier
  • Dihydridotetrakis(triphenylphosphine)ruthenium(II)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.