missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Dihydralazine sulfate

Product Code. 17221156 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17221156 250 mg 250mg
17211156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17221156 Supplier Thermo Scientific Chemicals Supplier No. 466232500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7327-87-9
Molecular Formula C8H12N6O4S

Specifications

Quantity 250 mg
Formula Weight 288.28
InChI Key BWHAMWGGORIDBK-UHFFFAOYSA-N
SMILES OS(O)(=O)=O.NNC1=C2C=CC=CC2=C(NN)N=N1
IUPAC Name 1,4-dihydrazinylphthalazine; sulfuric acid
Molecular Weight (g/mol) 288.28
Chemical Name or Material Dihydralazine sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.