missing translation for 'onlineSavingsMsg'
Learn More

Dihexylphthalate-D4, TRC

Product Code. 30340609
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30340609 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30340609 Supplier Toronto Research Chemicals Supplier No. TRCB185432100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid, bis-hexyl ester D4
CAS 1015854-55-3
Purity Grade Notes HPLC
Molecular Formula C20 D4 H26 O4
Quantity 100 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dihexyl ester, Phthalic Acid Di-n-hexyl Ester (3,4,5,6) D4, Phthalic acid, bis-hexyl ester D4, Phthalic Acid, Dihexyl Ester D4 (phenyl D4)
InChI Formula InChI=1S/C20H30O4/c1-3-5-7-11-15-23-19(21)17-13-9-10-14-18(17)20(22)24-16-12-8-6-4-2/h9-10,13-14H,3-8,11-12,15-16H2,1-2H3/i9D,10D,13D,14D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OCCCCCC)c(C(=O)OCCCCCC)c1[2H]
IUPAC Name dihexyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 338.4744
Formula Weight 338.2395
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.