missing translation for 'onlineSavingsMsg'
Learn More

Dihexylammonium acetate, 0.5 M solution in H2O, for ion pair chromatography, TRC

Product Code. 30387519
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387519 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387519 Supplier Toronto Research Chemicals Supplier No. TRCD44778810ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dihexylammonium acetate
CAS 366793-17-1
Molecular Formula C12H27N . C2H4O2
Quantity 10 mL
InChI Formula InChI=1S/C12H27N.C2H4O2/c1-3-5-7-9-11-13-12-10-8-6-4-2;1-2(3)4/h13H,3-12H2,1-2H3;1H3,(H,3,4)
SMILES CCCCCCNCCCCCC.CC(=O)O
IUPAC Name acetic acid;N-hexylhexan-1-amine
Molecular Weight (g/mol) 245.4
Formula Weight 245.2355
Concentration or Composition (by Analyte or Components) 0.5 M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.