missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexylammonium acetate, 0.5 M solution in H2O, for ion pair chromatography, TRC
Specifications
Specifications
| Chemical Name or Material | Dihexylammonium acetate |
| CAS | 366793-17-1 |
| Molecular Formula | C12H27N . C2H4O2 |
| Quantity | 50 mL |
| InChI Formula | InChI=1S/C12H27N.C2H4O2/c1-3-5-7-9-11-13-12-10-8-6-4-2;1-2(3)4/h13H,3-12H2,1-2H3;1H3,(H,3,4) |
| SMILES | CCCCCCNCCCCCC.CC(=O)O |
| IUPAC Name | acetic acid;N-hexylhexan-1-amine |
| Molecular Weight (g/mol) | 245.4 |
| Formula Weight | 245.2355 |
| Concentration or Composition (by Analyte or Components) | 0.5 M |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?