missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexadecyl Phosphate, TRC
Specifications
Specifications
| Chemical Name or Material | Dihexadecyl Phosphate |
| CAS | 2197-63-9 |
| Molecular Formula | C32H67O4P |
| Quantity | 1 g |
| Synonym | 1-Hexadecanol, hydrogen phosphate (8CI,9CI), Bis(hexadecyl) phosphate, CS 20A, Crodafos CS 20A, DAP 60H, Di(hexadecyl) monohydrogen phosphate, Di-n-hexadecyl phosphate, Dicetyl hydrogen phosphate, Dicetyl phosphate, Dihexadecyl hydrogen phosphate, Dihexadecyl phosphate, NSC 12429, Phosphoric acid dihexadecyl ester, 1-Hexadecanol, 1,1'-(hydrogen phosphate) |
| InChI Formula | InChI=1S/C32H67O4P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35-37(33,34)36-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,33,34) |
| SMILES | O=P(OCCCCCCCCCCCCCCCC)(O)OCCCCCCCCCCCCCCCC |
| IUPAC Name | dihexadecyl hydrogen phosphate |
| Molecular Weight (g/mol) | 546.85 |
| Formula Weight | 546.4777 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?