missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dihexadecyl(dimethyl)azanium Bromide, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Dihexadecyl(dimethyl)azanium Bromide |
| CAS | 70755-47-4 |
| Molecular Formula | C34H72BrN |
| Quantity | 500 mg |
| Synonym | 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, bromide (9CI), Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A, Dimethyldihexadecylammonium Bromide, N-hexadecyl-N,N-dimethyl-1-Hexadecanaminium bromide, Bis(hexadecyl) dimethyl ammonium bromide, Di-n-hexadecyldimethylammonium bromide, Dicetyldimethylammonium bromide, Dicetyldimonium bromide, Dihexadecyldimethylammonium bromide, Dimethyldihexadecylammonium bromide, Dimethyldipalmitylammonium bromide, Dipalmityldimethylammonium bromide, Konstanta A |
| InChI Formula | InChI=1S/C34H72N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(3,4)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-34H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC |
| IUPAC Name | dihexadecyl(dimethyl)azanium;bromide |
| Molecular Weight (g/mol) | 574.85 |
| Formula Weight | 573.4848 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction