missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin Monoclonal Antibody (DG D8-20), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110810
This item is not returnable.
View return policy
Description
MA1-10810 detects Digoxin. MA1-10810 has been successfully used in ELISA applications. The MA1-10810 immunogen is Digoxin, conjugated to KLH.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
Specifications
| Digoxin | |
| Monoclonal | |
| 1.46 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA | |
| DG D8-20 | |
| Unconjugated | |
| C41H64O14; DGX; Lanoxin | |
| Digoxin, conjugated to KLH. | |
| 500 μg | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction