missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin Monoclonal Antibody (DG D46), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110811
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-10811 detects Digoxin. MA1-10811 has been successfully used in ELISA applications. The MA1-10811 immunogen is Digoxin, conjugated to KLH.
Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.Specifications
Digoxin | |
Monoclonal | |
0.60 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA | |
DG D46 | |
Unconjugated | |
C41H64O14; DGX; Lanoxin | |
Digoxin, conjugated to KLH. | |
500 μg | |
Primary | |
Antibody | |
IgG1 |