missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Digoxin Monoclonal Antibody (38G12)

Product Code. 12847972
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12847972 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12847972 Supplier Invitrogen™ Supplier No. MID0302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MID0302 targets Digoxin in Western Blot, ELISA, IP and RIA applications and shows reactivity with labeled oligonucleotides and chemical samples. MID0302 was formerly sold as a Seradyn product.

Digoxin is a beta blocker drug used to treat heart problems, such as heart failure, high blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Digoxin
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 38G12
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C41H64O14; DGX; Lanoxin
Host Species Mouse
Immunogen Digoxin-BSA.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.