missing translation for 'onlineSavingsMsg'
Learn More

Digoxin, Lipomed

Product Code. 30462813
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462813 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462813 Supplier Lipomed Supplier No. LPMDOX192110
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Digoxin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Digoxin
CAS 20830-75-5
Analyte or Component Names Digoxin
Molecular Formula C41 H64 O14
Synonym Digoxin (8 CI), Cardigox, Cardiogoxin, Cardioxin, Cardixin, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES O[C@@]12 [C@@](C)([C@H](CC1)C3=CC(=O)OC3)[C@H](O)C[C@]4([C@]2(CC[C@]5([C@]4(C)CC[C@H](O[C@H]6 C[C@H](O)[C@H](O[C@H]7 C[C@H](O)[C@H](O[C@H]8 C[C@H](O)[C@H](O)[C@@H](C)O8)[C@@H](C)O7)[C@@H](C)O6)C5)[H])[H])[H]
IUPAC Name 3-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S,17 R)-3-[(2 R,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2 H-furan-5-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 780.94
Quantity 10 mg
Formula Weight 780.4296 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.