missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin Antibody (38G12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-78618-0.025mg
This item is not returnable.
View return policy
Description
Digoxin Monoclonal specifically detects Digoxin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Radioimmunoassay, Chemiluminescence Immunoassay.
Specifications
| Digoxin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, Chemiluminescence Immunoassay | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Chemical. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Chemiluminescence Immunoassay (CIA) | |
| 38G12 | |
| Unconjugated | |
| Lanoxin | |
| Digoxin-BSA | |
| 0.025 mg | |
| Primary | |
| Other | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction