missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin Antibody (26G10), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00
Specifications
| Antigen | Digoxin |
|---|---|
| Clone | 26G10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, Chemiluminescence Immunoassay |
| Applications | Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Chemiluminescence Immunoassay (CIA) |
Description
Digoxin Monoclonal specifically detects Digoxin in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Radioimmunoassay, Chemiluminescence Immunoassay.Specifications
| Digoxin | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Chemiluminescence Immunoassay (CIA) | |
| Unconjugated | |
| RUO | |
| Lanoxin | |
| IgG1 | |
| Affinity Purified | |
| Chemical. |
| 26G10 | |
| Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, Chemiluminescence Immunoassay | |
| Monoclonal | |
| Mouse | |
| Other | |
| Digoxin-BSA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title