missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxin, 1mg/ml in Methanol, Lipomed
Descripción
Digoxin, 1mg/ml in Methanol
Especificaciones
Especificaciones
| Chemical Name or Material | Digoxin |
| CAS | 20830-75-5 |
| Analyte or Component Names | Digoxin |
| Molecular Formula | C41 H64 O14 |
| Concentration | 1 mg/mL |
| Synonym | Digoxin (8 CI), Cardigox, Cardiogoxin, Cardioxin, Cardixin, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one, 4-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S)-3-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-5-[(2 S,4 S,5 R,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyl- oxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-5 H-furan-2-one |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | O[C@@]12 [C@@](C)([C@H](CC1)C3=CC(=O)OC3)[C@H](O)C[C@]4([C@]2(CC[C@]5([C@]4(C)CC[C@H](O[C@H]6 C[C@H](O)[C@H](O[C@H]7 C[C@H](O)[C@H](O[C@H]8 C[C@H](O)[C@H](O)[C@@H](C)O8)[C@@H](C)O7)[C@@H](C)O6)C5)[H])[H])[H] |
| IUPAC Name | 3-[(3 S,5 R,8 R,9 S,10 S,12 R,13 S,14 S,17 R)-3-[(2 R,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-5-[(2 S,4 S,5 S,6 R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-4-hydroxy-6-methyloxan-2-yl]oxy-12,14-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2 H-furan-5-one |
| Solvent or Matrix | Methanol |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?