missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Digoxigenin Recombinant Rabbit Monoclonal Antibody (PSH04-39)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA556506

Product Code. 30274798

  • £434.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Digoxigenin Recombinant Monoclonal Antibody for Western Blot, IHC (F), IHC (P), IP, ELISA, DB

Digoxin, also known as digitalis, is a purified cardiac glycoside extracted from the foxglove plant, Digitalis lanata. Its corresponding aglycone is digoxigenin, and its acetyl derivative is acetyldigoxin. Digoxin is widely used in the treatment of various heart conditions, namely atrial fibrillation, atrial flutter and sometimes heart failure that cannot be controlled by other medication. Digoxin preparations are commonly marketed under the trade names Lanoxin, Digitek, and Lanoxicaps. Digoxin is also used as an affinity system for nonradioactive labeling and detection of proteins and nucleic acids.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Digoxigenin
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Rabbit
Protein A
RUO
Chemical
Antibody
IgG
Dot Blot, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
PSH04-39
Unconjugated
C23H34O5; DIG; Digitalis
DIG-OVA.
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.