missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digoxigenin Antibody (PSH04-39), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32266
This item is not returnable.
View return policy
Description
Digoxigenin Monoclonal antibody specifically detects Digoxigenin in Nonspecies specific samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Dot Blot
Specifications
| Digoxigenin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, ELISA 1:5000-1:20000, Immunohistochemistry, Immunoprecipitation 1-2ug/sample, Immunohistochemistry-Paraffin 1:200-1:5000, Immunohistochemistry-Frozen 1:50-1:1000, Dot Blot 1:2000 | |
| DIG | |
| DIG-OVA | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| PSH04-39 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction