missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diglycolic anhydride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Diglycolic anhydride |
| CAS | 4480-83-5 |
| Molecular Formula | C4 H4 O4 |
| Quantity | 500 mg |
| Synonym | 3-Oxaglutaric anhydride, Diglycolic acid anhydride, Oxydiacetic anhydride, p-Dioxane-2,6-dione, 1,4-Dioxane-2,6-dione |
| InChI Formula | InChI=1S/C4H4O4/c5-3-1-7-2-4(6)8-3/h1-2H2 |
| SMILES | O=C1COCC(=O)O1 |
| IUPAC Name | 1,4-dioxane-2,6-dione |
| Molecular Weight (g/mol) | 116.072 |
| Formula Weight | 116.011 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction