missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic Acid, TRC

Product Code. 30374585
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30374585 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374585 Supplier Toronto Research Chemicals Supplier No. TRCD4463305G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diglycolic Acid
CAS 110-99-6
Molecular Formula C4H6O5
Quantity 5 g
Synonym 2,2'-Oxybis-Acetic Acid, Oxydi-acetic Acid, Diglycolic Acid, 2,2'-Oxydiacetic Acid, 3-Oxapentane-1,5-dioic Acid, 3-Oxapentanedioic Acid, Bis(carboxymethyl) Ether, NSC 8651, Oxodiacetic Acid, Oxybisacetic Acid, Oxydiacetic Acid, Oxydiethanolic Acid
InChI Formula InChI=1S/C4H6O5/c5-3(6)1-9-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
SMILES OC(=O)COCC(=O)O
Molecular Weight (g/mol) 134.09
Formula Weight 134.0215
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.