missing translation for 'onlineSavingsMsg'
Learn More

Diglycolic acid, 98%

Product Code. 10096003 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10096003 2.5 kg Plastic bottle 2.5kg
10295870 100 g Plastic bottle 100g
10544101 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096003 Supplier Thermo Scientific Acros Supplier No. 162260025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-99-6 | C4H6O5 | 134.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-99-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004309
InChI Key QEVGZEDELICMKH-UHFFFAOYSA-N
Synonym diglycolic acid, 2,2'-oxydiacetic acid, oxydiacetic acid, acetic acid, 2,2'-oxybis, oxybisacetic acid, oxydiethanolic acid, diglycolicacid, bis carboxymethyl ether, oxodiacetic acid, 3-oxapentanedioic acid
PubChem CID 8088
IUPAC Name 2-(carboxymethoxy)acetic acid
SMILES OC(=O)COCC(O)=O

Specifications

Melting Point 141.0°C to 145.0°C
Color White to Beige
Assay Percent Range 98%
Linear Formula O(CH2CO2H)2
Beilstein 03, 234
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 134.09
Percent Purity 98%
Quantity 2.5 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Crystalline Powder
Chemical Name or Material Diglycolic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Diglycolic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.