missing translation for 'onlineSavingsMsg'
Learn More

Diglycidyl Resorcinol Ether, TRC

Product Code. 30385881
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385881 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385881 Supplier Toronto Research Chemicals Supplier No. TRCD44634310ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Resorcinol diglycidyl ether
CAS 101-90-6
Molecular Formula C12 H14 O4
Quantity 10 mL
Synonym Oxirane, 2,2'-[1,3-phenylenebis(oxymethylene)]bis-, Benzene, m-bis(2,3-epoxypropoxy)- (6CI,7CI,8CI), 2,2'-[1,3-Phenylenebis(oxymethylene)]bis[oxirane], 1,3-Bis(2,3-epoxypropoxy)benzene, 1,3-Diglycidyloxybenzene, 1,3-Dihydroxybenzene diglycidyl ether, Diglycidyl resorcinol ether, NSC 76621, Resorcinol bis(2,3-epoxypropyl) ether, Resorcinol diglycidyl ether, Resorcinol glycidyl ether, m-Bis(2,3-epoxypropoxy)benzene, m-Bis(glycidyloxy)benzene
InChI Formula InChI=1S/C12H14O4/c1-2-9(13-5-11-7-15-11)4-10(3-1)14-6-12-8-16-12/h1-4,11-12H,5-8H2
SMILES C(Oc1cccc(OCC2CO2)c1)C3CO3
IUPAC Name 2-[[3-(oxiran-2-ylmethoxy)phenoxy]methyl]oxirane
Molecular Weight (g/mol) 222.24
Formula Weight 222.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.