missing translation for 'onlineSavingsMsg'
Learn More

Digitoxigenin Mono-Beta-D-digitoxoside, TRC

Product Code. 30386008
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386008 5 mg -
30387109 1 mg 1mg
30381342 0.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386008 Supplier Toronto Research Chemicals Supplier No. TRCD4508355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18404-43-8
Molecular Formula C29H44O7
Molecular Weight (g/mol) 504.66
Synonym 3β-[(2,6-Dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxy-5β-card-20(22)-enolide, Evatromonoside, (3β,5β)-3-[(2,6-Dideoxy-β-D-ribo-hexopyranosyl)oxy]-14-hydroxycard-20(22)-enolide, 2,6-Dideoxy-β-D-ribo-hexopyranoside-digitoxigenin-3, 2,6-Dideoxy-β-D-ribo-hexopyranoside Digitoxigenin, Digitoxigenin Digitoxoside Digitoxigenin Monodigitoxoside
SMILES C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2CC[C@@]3(C)[C@H](CC[C@@H]4[C@@H]3CC[C@]5(C)[C@H](CC[C@]45O)C6=CC(=O)OC6)C2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H44O7/c1-16-26(32)23(30)14-25(35-16)36-19-6-9-27(2)18(13-19)4-5-22-21(27)7-10-28(3)20(8-11-29(22,28)33)17-12-24(31)34-15-17/h12,16,18-23,25-26,30,32-33H,4-11,13-15H2,1-3H3/t16-,18-,19+,20-,21+,22-,23+,25+,26-,27+,28-,29+/m1/s1
Chemical Name or Material Digitoxigenin Mono-Beta-D-digitoxoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.