missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digitoxigenin-21,23,23-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Digitoxigenin-21,23,23-d3 |
| Molecular Formula | C23H31D3O4 |
| Quantity | 1 mg |
| Synonym | (3β,5β)-3,14-Dihydroxy-card-20(22)-enolide, 3β,14-Dihydroxy-5β-card-20(22)-enolide, (+)-Digitoxigenin, Cerberigenin, NSC 407806, Δ20:22-3,14,21-Trihydroxynorcholenic Acid Lactone |
| InChI Formula | InChI=1S/C23H34O4/c1-21-8-5-16(24)12-15(21)3-4-19-18(21)6-9-22(2)17(7-10-23(19,22)26)14-11-20(25)27-13-14/h11,15-19,24,26H,3-10,12-13H2,1-2H3/t15-,16+,17-,18+,19-,21+,22-,23+/m1/s1/i11D,13D2 |
| SMILES | [H][C@@]12CC[C@]3([H])C[C@@H](O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@]2(O)CC[C@@H]4C5=C([2H])C(OC5([2H])[2H])=O |
| Molecular Weight (g/mol) | 377.53 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?