missing translation for 'onlineSavingsMsg'
Learn More

Digitonin, MedChemExpress

Product Code. 16493956
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493956 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493956 Supplier Medchem Express Supplier No. HYN4000
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Digitonin, a glycoside obtained from Digitalis purpurea, could increase cell permeability by binding to cholesterol molecules and reduce tumor growth. Digitonin is an natural detergent.

Digitonin, a glycoside obtained from Digitalis purpurea, could increase cell permeability by binding to cholesterol molecules and reduce tumor growth. Digitonin is an natural detergent.

Specifications

Chemical Name or Material Digitonin
CAS 11024-24-1
Color Light Nude
Purity Grade Notes Research
Molecular Formula C56H92O29
Quantity 25 mg
Solubility Information H2O : 100 mg/mL (81.35 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (81.35 mM; Need ultrasonic)
SMILES C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@@H](C)CO4)([H])[C@@H](O)[C@@]1([H])[C@@](CC[C@]5([H])[C@@]6(C[C@@H](O)[C@H](O[C@]([C@@H]([C@@H](O)[C@H]7O[C@@](O[C@H](CO)[C@@H](O)[C@@H]8O[C@@](OC[C@@H](O)[C@@H]9O)([H])[C@@H]9O)([H])[C@@H]8O[C@@](O[C@H](CO)[C@H](O)[C@@H]%10O[C@]([C@@H]([C@@H](O)[C@@H]%11O)O)([H])O[C@@H]%11CO)([H])[C@@H]%10O)O)([H])O[C@@H]7CO)C5)C)([H])[C@]6([H])CC2
Molecular Weight (g/mol) 1229.31
Formula Weight 1229.31
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.