Learn More
WTW™ Digital Triple Channel Multi Parameter Sonde Kit for WTW™ IDS sensors
Click to view available options
For Use With (Equipment):
IDS Sensors, SensoLyt 900 -P, TetraCon 925 -P, FDO 925 -P
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Designed for depth measurements to 100m and for pH, conductivity, DO measurement |
| Includes | Built-in pressure sensor, guard, weight, protection cap, instruction manual, CD- ROM with software, 1.5m connection cable of AS IDS/1.5, carry case including SensoLyt™ 900 -P, TetraCon™ 925 -P |
| For Use With (Equipment) | IDS Sensors, SensoLyt 900 -P, TetraCon 925 -P, FDO 925 -P |