missing translation for 'onlineSavingsMsg'
Learn More

TFA Dostmann Digital Timer and Stopwatch

Product Code. 17963111
Change view
Click to view available options
Display:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display unitSize
17963111 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17963111 Supplier TFA Dostmann Supplier No. 38.2038.02
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This digital timer and stopwatch is the perfect everyday companion. Thanks to the extra loud alarm, you are reliably reminded. The buttons under the display allow you to set the right time quickly and easily.

This digital timer with stopwatch is the perfect everyday companion. It provides you with the perfect breakfast egg, times your training sessions or helps you with time management at work. The buttons under the display allow you to set the right time quickly and easily. Thanks to the extra loud alarm, you are reliably reminded. The timer can be attached in a variety of ways: with a magnet, stand or hanger.

  • Extra-loud alarm
  • Quick and easy time setting
  • With magnet, table stand and hanging loop
  • Large, easy-to-read display
  • Max. time setting: 99 min, 50 sec

Specifications

Product Type Digital Timer
Time Range 99 min. 59 sec.
Alarms Audible
Display LCD
Battery Type 1 x 3 V CR2032
Color Black
Weight (Metric) 66 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.