missing translation for 'onlineSavingsMsg'
Learn More

TFA Dostmann Digital Timer and Stopwatch with Aluminium Surface

Product Code. 17971312
Change view
Click to view available options
Display:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display unitSize
17971312 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971312 Supplier TFA Dostmann Supplier No. 38.2013.54
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This digital timer and stopwatch with aluminium surface is your perfect everyday companion. It can provide you with the perfect breakfast egg, time your training sessions or help you with time management at work. Its large display is framed by a high-quality aluminium front and is easy to read.

This digital timer with stopwatch is your perfect everyday companion. It can provide you with the perfect breakfast egg, time your training sessions or help you with time management at work. Its large display is framed by a high-quality aluminium front and is easy to read. Thanks to its practical magnet, you can quickly and easily attach the timer to metal surfaces or set it up using the stand.

  • Front made of high-grade aluminium
  • Large display
  • With mounting magnet and stand
  • Max. time setting: 99 min, 59 sec

Specifications

Product Type Digital Timer
Time Range 99 min. 59 sec.
Alarms Audible
Display LCD
Battery Type 1 x 1.5 V AAA
Color Silver
Weight (Metric) 70 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.