missing translation for 'onlineSavingsMsg'
Learn More

TFA Dostmann Digital Timer and Stopwatch 99 Hours

Product Code. 17931312
Change view
Click to view available options
Display:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display unitSize
17931312 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17931312 Supplier TFA Dostmann Supplier No. 38.2049.02
Add to basket
Add to basket
This item is not returnable. View return policy

The digital timer and stopwatch 99HOURS has a memory function and displays three preset times. You can also interrupt your timer and quickly reactivate it with just one press. The pre-alarm for example, if you set it for one minute, an alarm sounds one minute before the time expires.

As a digital timer, the 99HOURS is the solution. With the 99HOURS, you have three preset times right on your screen. You can quickly reactivate it with just one press. The pre-alarm may also be useful in certain situations. You can set it according to your needs. For example, if you set it for one minute, an alarm sounds one minute before the time expires. Perfect for those who like to be prepared in good time. This timer has two volume levels to choose from or you can switch off the sound completely. Then, only the LED warning light lights up as a visual alarm. If the timer is not stopped, it displays the time elapsed. With a maximum running time of 99 hours and 59 minutes, this timer covers a time span of over four days to one second. Few buttons, many functions, robust plastic housing. Equipped with a magnet, stand and hanging loop, you can always find a handy place for it. The only other thing you need are two AAA batteries.

  • Count-up and count-down timer
  • Memory function (3 times)
  • 3 volume levels (loud, normal, off)
  • LED alarm light
  • Elapsed time display
  • Pre-alarm (freely adjustable)
  • Hours/minutes and minutes/seconds display
  • Up to 99 h/59 min or 99 min/59 s
  • With magnet for fixing, hanging slot or table stand

Specifications

Product Type Digital Timer
Time Range 99 hr. 59 min. 59 sec.
Alarms Audible
Display LCD
Battery Type 2 x 1.5 V AAA
Color White
Weight (Metric) 70 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.