missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Digital Manometer

Product Code. 15237948
Encompass_Preferred
Change view
Click to view available options
Pressure Range:
30 psi
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pressure Range Unit Size
15237948 30 psi Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15237948 Supplier Cole-Parmer™ Supplier No. 6806276
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE Compliance
For Use With (Application) Measures positive, negative and differential pressures
Size (Metric) Fitting Dual 3.17 mm NPT(F)
Accuracy ±0.5% Full-scale at 60 to 78°F; ±1.5% Full-scale from 32 to 60°F and 78 to 104
Display Type LCD
Pressure Range 30 psi
Product Type Digital Manometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.