missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Digital Dry Block Heater and Cooler. -3°C to +150°C, UK

Product Code. 17755009
Change view
Click to view available options
No. of Blocks:
2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Blocks Unit Size
17755009 2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17755009 Supplier Grant Instruments™ Supplier No. CH3150 UK
  • ONLINE EXCLUSIVE PRICE
    £1795.00
    £1757.00 / Pack of 1
    Save £38.00
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Grant-bio CH3-150 Combitherm-2 dry block heating cooling system is specially designed to thermostabilise materials at temperatures from –3°C to +150°C according to methods of analysis.

The Grant-bio CH3-150 Combitherm-2 dry block heating cooling system is specially designed to thermostabilise materials at temperatures from –3°C to +150°C according to methods of analysis.
  • Stability: ±0.1ºC
  • Independently regulated heating and cooling blocks in the same unit
  • Digital timer with sound alarm: 1 min to 99 h 59 min (increment 1 min)
  • User adjustable programmes (temperature and time): 16 (heating) + 16 (cooling)
  • Digital temperature control for optimum precision
  • Temperature Calibration Function
  • Protective lid

The CH3-150 has seven interchangeable blocks available. Custom blocks available on request.

Applications:

Life-science – storing restriction enzymes, nick translation, ligation reactions, restriction digests, protein solubilisation for PAGE, warm incubation of microcentrifuge tubes for hybridisation, enzyme reactions and deactivations

Clinical – cooling blood samples prior to coagulation testing. acylcarnitines derivatisation, MRSA and PBP2 latex testing, heating flush/media used in egg recovery, fertility to keep test tubes at correct temperature during egg collection

Incubating samples at set temperatures, heating block for boiling of solutions in tubes

Life-science-cell digestion, DNA/RNA extraction, post sequencing PCT clean-up-dry down step, boiling in vitro DNA RNA/ protein samples, incubating invitro reactions/digestions, extraction of DNA for real-time PCR analysis, denaturing nucleic acid and protein samples

Industrial – digestion of environmental samples for chemical oxygen demand analysis, soil digests, maintaining temperatures.

TRUSTED_SUSTAINABILITY

Specifications

Color White
Description Digital dry block heater and cooler
Holds Blocks B2-50, B10-13, B10-16, B5-29, B6-25, B18-12 and B23-1.5
Plug Type UK
Temperature Range (Metric) -3°C to +20°C
Type Digital Dry Block Heater and Cooler
Display Yes
No. of Blocks 2
Temperature Stability ±0.1°C
Timer 1 min. to 99 hrs 59 min. (1 min.increments)
Warranty 2 years
Length (Metric) 43 cm
Width (Metric) 295 mm
Height (Metric) 35 cm
Depth (Metric) 285 mm
Shipping Weight (Metric) 7 kg
Weight (Metric) 5.6 kg
Voltage 230 V
Wattage 430 W
Amperage 1.8 A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.