Learn More
Gilson™ Digital Dry Bath Block for NMR tubes
Shop All Gilson Products
Click to view available options
Unit Size:
Each
Description
Accessory for Digital Dry Bath Series.
Specifications
Specifications
| For Use With (Equipment) | Gilson Digital Dry Bath Series |
| Holds | 30 x NMR tubes |
| Type | Dry Bath Block |