Learn More
Leica Microsystems EC3 Digital Camera
Description
- Fits to a trinocular tube on top of the microscope
- Software to allow live image preview and image capture included
- Capture high-resolution color images of 3.1 megapixels
- Powerful camera controls include shading correction and optional predefined illumination settings.
- Create high quality images for use in PowerPoint™ presentations, lab reports, and course related material
- Images can be posted on websites or intranets for easy sharing
- Single camera cable connects to USB2 port
- Archive standard and unusual specimens
- Easy connection to all c-mount- and video adapters
- Use with external projectors
Specifications
Specifications
| For Use With (Equipment) | DM500 and DM750 microscopes |