Learn More
Key Industrial Equipment™ Digital caliper
Brand:  Key Industrial Equipment™ A343618
Encompass
Product Code. 12818719
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Supplied in plastic storage case. Extra large display reading to 0. 01mm and 0. 0005 in.
  • Measuring range 0-150mm
  • SR44 battery included