missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Digest-All 3 (Pepsin)
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
This product may be used to digest formalin-fixed, paraffin-embedded tissues (see Digest-All Kit above), and will improve the signal of certain antigens. It contains 50 mL of ready-to-use Pepsin. Digest-All™ 3 is stable for 12 months.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C). Shelf life: 18 months from date of manufacture |
| Type | Antigen Retrieval Reagent |
| Product Line | Digest-All |
| Quantity | 50 mL |
| Reagent Type | Antigen Retrieval Reagent |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction