missing translation for 'onlineSavingsMsg'
Learn More

Digeranyl bisphosphonate, MedChemExpress

Product Code. 16462398
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462398 5 mg 5mg
16460199 10 mg 10mg
16425437 50 mg 50mg
16403189 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462398 Supplier Medchem Express Supplier No. HYU00145
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Digeranyl bisphosphonate (DGBP) is a potent geranylgeranylpyrophosphate (GGPP) synthase inhibitor, which inhibits geranylgeranylation of Rac1.

Digeranyl bisphosphonate (DGBP) is a potent geranylgeranylpyrophosphate (GGPP) synthase inhibitor, which inhibits geranylgeranylation of Rac1.

Specifications

Chemical Name or Material Digeranyl bisphosphonate
CAS 878143-03-4
Color White
Purity Grade Notes Research
Molecular Formula C21H34Na4O6P2
Quantity 5 mg
Synonym DGBP
Solubility Information H2O : 2 mg/mL (3.73 mM; Need ultrasonic) ∣DMSO : 2 mg/mL (3.73 mM; Need ultrasonic)
SMILES C/C(C)=C\CC/C(C)=C/CC(P(O[Na])(O[Na])=O)(P(O[Na])(O[Na])=O)C/C=C(C)/CC/C=C(C)\C
Molecular Weight (g/mol) 536.4
Formula Weight 536.4
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.