missing translation for 'onlineSavingsMsg'
Learn More

Difuroyl Disulfide, TRC

Product Code. 30383351
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383351 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383351 Supplier Toronto Research Chemicals Supplier No. TRCD59056025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Difuroyl Disulfide
CAS 51278-29-6
Molecular Formula C10 H6 O4 S2
Quantity 25 mg
Synonym Furan-2-carboxylic Dithioperoxyanhydride, Bis(2-furanylcarbonyl) Disulfide
InChI Formula InChI=1S/C10H6O4S2/c11-9(7-3-1-5-13-7)15-16-10(12)8-4-2-6-14-8/h1-6H
SMILES O=C(SSC(=O)c1occc1)c2occc2
IUPAC Name S-(furan-2-carbonylsulfanyl) furan-2-carbothioate
Molecular Weight (g/mol) 254.282
Formula Weight 253.971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.