missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Difluprednate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Difluprednate is a topical corticosteroid, which is thought to act by the induction of phospholipase A2 inhibitory proteins (lipocortins). Difluprednate is used for the symptomatic treatment of inflammation and pain associated with ocular surgery.
Specifications
Specifications
| Chemical Name or Material | Difluprednate |
| CAS | 23674-86-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H34F2O7 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 250 mg/mL (491.59 mM; Need ultrasonic) |
| SMILES | O=C([C@]([C@]12C)(CC[C@@]1([H])[C@@](C[C@H](F)C3=CC4=O)([H])[C@@](F)([C@]3(C=C4)C)[C@@H](O)C2)OC(CCC)=O)COC(C)=O |
| Molecular Weight (g/mol) | 508.55 |
| Formula Weight | 508.55 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction