missing translation for 'onlineSavingsMsg'
Learn More

Diflunisal, TRC

Product Code. 30133250
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133250 1 g 1g
30132854 25 g 25g
30006936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133250 Supplier Toronto Research Chemicals Supplier No. TRCD4457511G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22494-42-4
Molecular Formula C13 H8 F2 O3
Molecular Weight (g/mol) 250.2
Synonym 3-Biphenylcarboxylic acid, 2',4'-difluoro-4-hydroxy-, 2',4'-Difluoro-4-hydroxy-1,1'-biphenyl-3-carboxylic acid, 2',4'-Difluoro-4-hydroxybiphenyl-3-carboxylic acid, 5-(2,4-Difluorophenyl)salicylic acid, Adomal, Difludol, Diflunisal, Diflusinal, Dolisal, Doloban, Dolobid, Dolobis, Flovacil, Fluniget, Fluodonil, Flustar, MK 647
IUPAC Name 5-(2,4-difluorophenyl)-2-hydroxybenzoic acid
SMILES OC(=O)c1cc(ccc1O)c2ccc(F)cc2F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H8F2O3/c14-8-2-3-9(11(15)6-8)7-1-4-12(16)10(5-7)13(17)18/h1-6,16H,(H,17,18)
Formula Weight 250.0442
Chemical Name or Material Diflunisal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.